[4-(1H-Imidazol-2-yl)phenyl]amine dihydrochloride

Supplier: Matrix Scientific

077577-500MG 077577-1G 077577-5G
102666-958EA 646.15 USD
102666-958 102666-960 102666-962
[4-(1H-Imidazol-2-yl)phenyl]amine dihydrochloride
[4-(1H-Imidazol-2-yl)phenyl]amine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₁Cl₂N₃
MDL Number: MFCD16040008
CAS Number: 1235440-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR