2-(4-Nitrophenyl)-1H-imidazole hydrochloride
Supplier: Matrix Scientific
077576-500MG
077576-1G
102666-954EA
387.69
USD
102666-954
102666-956
2-(4-Nitrophenyl)-1H-imidazole hydrochloride
2-(4-Nitrophenyl)-1H-imidazole hydrochloride
Hazard: Irritant
Formula:
C₉H₈ClN₃O₂ MW: 225.64 g/mol |
MDL Number:
MFCD12912834 CAS Number: 1193389-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...