tert-Butyl-4-(2-hydrazino-2-oxoethyl)piperazine-1-carboxylate

Supplier: Matrix Scientific

077572-500MG 077572-1G
102666-932EA 492.31 USD
102666-932 102666-934
tert-Butyl-4-(2-hydrazino-2-oxoethyl)piperazine-1-carboxylate
tert-Butyl-4-(2-hydrazino-2-oxoethyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₂N₄O₃
MW: 258.32 g/mol
MDL Number: MFCD06795935
CAS Number: 1338673-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR