tert-Butyl {2-[3-(benzoylamino)-1H-1,2,4-triazol-5-yl]ethyl}carbamate
Supplier: Matrix Scientific
077568-500MG
077568-1G
077568-5G
102666-916EA
193.85
USD
102666-916
102666-918
102666-920
tert-Butyl {2-[3-(benzoylamino)-1H-1,2,4-triazol-5-yl]ethyl}carbamate
tert-Butyl {2-[3-(benzoylamino)-1H-1,2,4-triazol-5-yl]ethyl}carbamate
Hazard: Irritant
Formula:
C₁₆H₂₁N₅O₃ |
MDL Number:
MFCD20441377 CAS Number: 1338494-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...