5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride

Supplier: Matrix Scientific

077567-500MG 077567-1G 077567-5G
102666-910EA 646.15 USD
102666-910 102666-912 102666-914
5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride
5-(2-Aminoethyl)-1H-1,2,4-triazol-3-amine dihydrochloride

Hazard: Irritant

Formula: C₄H₁₁Cl₂N₅
MW: 200.07 g/mol
MDL Number: MFCD20441445
CAS Number: 99839-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR