2-(Azidomethyl)[1,2,4]triazolo[1,5-a]pyrimidine
Supplier: Matrix Scientific
077566-500MG
077566-1G
102666-906EA
646.15
USD
102666-906
102666-908
2-(Azidomethyl)[1,2,4]triazolo[1,5-a]pyrimidine
2-(Azidomethyl)[1,2,4]triazolo[1,5-a]pyrimidine
Hazard: Irritant
Formula:
C₆H₅N₇ |
MDL Number:
MFCD20441376 CAS Number: 1338494-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...