2-(Chloromethyl)[1,2,4]triazolo[1,5-a]pyrimidine

Supplier: Matrix Scientific

077565-500MG 077565-1G 077565-5G
102666-900EA 646.15 USD
102666-900 102666-902 102666-904
2-(Chloromethyl)[1,2,4]triazolo[1,5-a]pyrimidine
2-(Chloromethyl)[1,2,4]triazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₆H₅ClN₄
MDL Number: MFCD20441375
CAS Number: 1338495-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR