5-Fluoro-6-methoxypyrimidin-2(1H)-one

Supplier: Matrix Scientific

077564-500MG 077564-1G
102666-896EA 646.15 USD
102666-896 102666-898
5-Fluoro-6-methoxypyrimidin-2(1H)-one
5-Fluoro-6-methoxypyrimidin-2(1H)-one

Hazard: Irritant

Formula: C₅H₅FN₂O₂
MDL Number: MFCD20441374
CAS Number: 22462-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR