6-(Dimethylamino)-5-fluoropyrimidin-2(1H)-one
Supplier: Matrix Scientific
077562-500MG
077562-1G
102666-890EA
516.92
USD
102666-890
102666-892
6-(Dimethylamino)-5-fluoropyrimidin-2(1H)-one
6-(Dimethylamino)-5-fluoropyrimidin-2(1H)-one
Hazard: Irritant
Formula:
C₆H₈FN₃O |
MDL Number:
MFCD20441373 CAS Number: 1338494-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...