tert-Butyl-4-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Supplier: Matrix Scientific

077560-500MG 077560-1G 077560-5G
102666-878EA 646.15 USD
102666-878 102666-880 102666-882
tert-Butyl-4-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
tert-Butyl-4-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O₂
MW: 282.77 g/mol
MDL Number: MFCD20441456
CAS Number: 1401425-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR