tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
Supplier: Matrix Scientific
077559-500MG
077559-1G
077559-5G
102666-872EA
516.92
USD
102666-872
102666-874
102666-876
tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
tert-Butyl-6-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₉ClN₂O₂ MW: 282.77 g/mol |
MDL Number:
MFCD20441458 CAS Number: 1401425-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...