tert-Butyl-3-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Supplier: Matrix Scientific

077555-500MG 077555-1G 077555-5G
102666-852EA 516.92 USD
102666-852 102666-854 102666-856
tert-Butyl-3-(aminomethyl)-1H-indole-1-carboxylate hydrochloride
tert-Butyl-3-(aminomethyl)-1H-indole-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O₂
MW: 282.77 g/mol
MDL Number: MFCD20441455
CAS Number: 1401425-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR