(2E)-1-(Phenylsulfonyl)acetone oxime

Supplier: Matrix Scientific

077554-500MG 077554-1G
102666-848EA 387.69 USD
102666-848 102666-850
(2E)-1-(Phenylsulfonyl)acetone oxime
(2E)-1-(Phenylsulfonyl)acetone oxime

Hazard: Irritant

Formula: C₉H₁₁NO₃S
MDL Number: MFCD01924614
CAS Number: 343348-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR