1-(1,3-Benzothiazol-2-yl)piperidine-2-carboxylic acid
Supplier: Matrix Scientific
077547-500MG
077547-1G
077547-5G
102666-810EA
387.69
USD
102666-810
102666-812
102666-814
1-(1,3-Benzothiazol-2-yl)piperidine-2-carboxylic acid
1-(1,3-Benzothiazol-2-yl)piperidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₂S |
MDL Number:
MFCD09739050 CAS Number: 1163289-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...