2-(1H-1,2,4-Triazol-5-yl)pyrazine

Supplier: Matrix Scientific

077545-500MG 077545-1G
102666-802EA 646.15 USD
102666-802 102666-804
2-(1H-1,2,4-Triazol-5-yl)pyrazine
2-(1H-1,2,4-Triazol-5-yl)pyrazine

Hazard: Irritant

Formula: C₆H₅N₅
MDL Number: MFCD20441369
CAS Number: 130612-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR