Dimethyl-2,2'-hydrazine-1,2-diyldiacetate hydrochloride

Supplier: Matrix Scientific

077542-500MG 077542-1G 077542-5G
102666-784EA 387.69 USD
102666-784 102666-786 102666-788
Dimethyl-2,2'-hydrazine-1,2-diyldiacetate hydrochloride
Dimethyl-2,2'-hydrazine-1,2-diyldiacetate hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O₄
MDL Number: MFCD20441453
CAS Number: 1401425-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR