2-(2-Methoxyethoxy)-4-methyl-1,3-thiazole-5-carboxylic acid

Supplier: Matrix Scientific

077531-500MG 077531-1G 077531-5G
102666-738EA 387.69 USD
102666-738 102666-740 102666-742
2-(2-Methoxyethoxy)-4-methyl-1,3-thiazole-5-carboxylic acid
2-(2-Methoxyethoxy)-4-methyl-1,3-thiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₁NO₄S
MDL Number: MFCD20441365
CAS Number: 1338494-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR