5-(4-Methoxyphenyl)[1,3]thiazolo-[4,3-b][1,3,4]thiadiazol-2-amine
Supplier: Matrix Scientific
077525-500MG
077525-1G
102666-712EA
516.92
USD
102666-712
102666-714
5-(4-Methoxyphenyl)[1,3]thiazolo-[4,3-b][1,3,4]thiadiazol-2-amine
5-(4-Methoxyphenyl)[1,3]thiazolo-[4,3-b][1,3,4]thiadiazol-2-amine
Hazard: Irritant
Formula:
C₁₁H₁₁N₃OS₂ |
MDL Number:
MFCD00807192 CAS Number: 313670-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...