5-Phenyl[1,3]thiazolo[4,3-b][1,3,4]thiadiazol-2-amine

Supplier: Matrix Scientific

077524-500MG 077524-1G 077524-5G
102666-706EA 516.92 USD
102666-706 102666-708 102666-710
5-Phenyl[1,3]thiazolo[4,3-b][1,3,4]thiadiazol-2-amine
5-Phenyl[1,3]thiazolo[4,3-b][1,3,4]thiadiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₉N₃S₂
MDL Number: MFCD00812922
CAS Number: 137609-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR