4-Amino-5-(3-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077523-500MG 077523-1G 077523-5G
102666-700EA 387.69 USD
102666-700 102666-702 102666-704
4-Amino-5-(3-methoxyphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(3-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₀N₄OS
MDL Number: MFCD01166533
CAS Number: 150093-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR