4-Amino-5-{[2-(trifluoromethyl)-1H-benzimidazol-1-yl]methyl}-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077520-500MG 077520-1G
102666-696EA 258.46 USD
102666-696 102666-698
4-Amino-5-{[2-(trifluoromethyl)-1H-benzimidazol-1-yl]methyl}-4H-1,2,4-triazole-3-thiol
4-Amino-5-{[2-(trifluoromethyl)-1H-benzimidazol-1-yl]methyl}-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₉F₃N₆S
MDL Number: MFCD20441362
CAS Number: 246163-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR