4-Amino-5-{[benzyl(methyl)amino]methyl}-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077519-500MG 077519-1G
102666-692EA 193.2 USD
102666-692 102666-694
4-Amino-5-{[benzyl(methyl)amino]methyl}-4H-1,2,4-triazole-3-thiol
4-Amino-5-{[benzyl(methyl)amino]methyl}-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₁H₁₅N₅S
MDL Number: MFCD18169600
CAS Number: 1207326-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR