4-Amino-5-[(3-phenylpyrrolidin-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077518-500MG
077518-1G
077518-5G
102666-686EA
387.69
USD
102666-686
102666-688
102666-690
4-Amino-5-[(3-phenylpyrrolidin-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(3-phenylpyrrolidin-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₁₃H₁₇N₅S |
MDL Number:
MFCD20441361 CAS Number: 1338494-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...