4-Amino-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077517-500MG 077517-1G
102666-682EA 386.4 USD
102666-682 102666-684
4-Amino-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₁₂N₆S
MDL Number: MFCD20441360
CAS Number: 1338494-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR