4-Amino-5-(azepan-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077516-500MG
077516-1G
077516-5G
102666-676EA
387.69
USD
102666-676
102666-678
102666-680
4-Amino-5-(azepan-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(azepan-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₉H₁₇N₅S |
MDL Number:
MFCD18203594 CAS Number: 1260923-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...