4-Amino-5-(morpholin-4-ylmethyl)-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077515-500MG
077515-1G
077515-5G
102666-670EA
387.69
USD
102666-670
102666-672
102666-674
4-Amino-5-(morpholin-4-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(morpholin-4-ylmethyl)-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₇H₁₃N₅OS |
MDL Number:
MFCD09860367 CAS Number: 155219-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...