4-Amino-5-(piperidin-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077513-500MG 077513-1G 077513-5G
102666-658EA 289.8 USD
102666-658 102666-660 102666-662
4-Amino-5-(piperidin-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(piperidin-1-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₁₅N₅S
MDL Number: MFCD13187435
CAS Number: 155219-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR