4-Amino-5-(1-naphthylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077510-500MG 077510-1G
102666-650EA 387.69 USD
102666-650 102666-652
4-Amino-5-(1-naphthylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(1-naphthylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₂N₄S
MW: 256.33 g/mol
MDL Number: MFCD03653126
CAS Number: 151297-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR