4-Amino-5-(3-isobutyl-1H-pyrazol-5-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077509-500MG 077509-1G 077509-5G
102666-644EA 516.92 USD
102666-644 102666-646 102666-648
4-Amino-5-(3-isobutyl-1H-pyrazol-5-yl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(3-isobutyl-1H-pyrazol-5-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₄N₆S
MDL Number: MFCD13370785
CAS Number: 1093613-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR