4-Amino-5-[(2-pyridin-2-yl-1H-benzimidazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077499-500MG 077499-1G 077499-5G
102666-590EA 193.85 USD
102666-590 102666-592 102666-594
4-Amino-5-[(2-pyridin-2-yl-1H-benzimidazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(2-pyridin-2-yl-1H-benzimidazol-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₃N₇S
MDL Number: MFCD08285769
CAS Number: 895835-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR