4-Amino-5-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077494-500MG
077494-1G
077494-5G
102666-564EA
387.69
USD
102666-564
102666-566
102666-568
4-Amino-5-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₅H₁₀N₄OS |
MDL Number:
MFCD10567686 CAS Number: 1017664-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...