4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077491-500MG
077491-1G
077491-5G
102666-546EA
387.69
USD
102666-546
102666-548
102666-550
4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(1H-1,2,3-benzotriazol-1-ylmethyl)-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₉H₉N₇S |
MDL Number:
MFCD04035853 CAS Number: 122503-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...