4-Amino-3-mercapto-6-(4-methylphenyl)-1,2,4-triazin-5(4H)-one
Supplier: Matrix Scientific
077490-500MG
077490-1G
077490-5G
102666-540EA
387.69
USD
102666-540
102666-542
102666-544
4-Amino-3-mercapto-6-(4-methylphenyl)-1,2,4-triazin-5(4H)-one
4-Amino-3-mercapto-6-(4-methylphenyl)-1,2,4-triazin-5(4H)-one
Hazard: Irritant
Formula:
C₁₀H₁₀N₄OS MW: 234.28 g/mol |
MDL Number:
MFCD11868885 CAS Number: 22445-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...