4-Amino-5-(2-methyl-3-furyl)-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077488-500MG
077488-1G
077488-5G
102666-528EA
387.69
USD
102666-528
102666-530
102666-532
4-Amino-5-(2-methyl-3-furyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-methyl-3-furyl)-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₇H₈N₄OS |
MDL Number:
MFCD04608597 CAS Number: 560998-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...