(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)(diphenyl)methanol
Supplier: Matrix Scientific
077486-500MG
077486-1G
077486-5G
102666-516EA
258.46
USD
102666-516
102666-518
102666-520
(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)(diphenyl)methanol
(4-Amino-5-mercapto-4H-1,2,4-triazol-3-yl)(diphenyl)methanol
Hazard: Irritant
Formula:
C₁₅H₁₄N₄OS |
MDL Number:
MFCD13370780 CAS Number: 175604-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...