4-Amino-5-(4-bromophenyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077484-500MG 077484-1G 077484-5G
102666-504EA 387.69 USD
102666-504 102666-506 102666-508
4-Amino-5-(4-bromophenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(4-bromophenyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₇BrN₄S
MDL Number: MFCD00806254
CAS Number: 85106-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR