4-Amino-5-[(4-fluorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077483-500MG 077483-1G 077483-5G
102666-498EA 387.69 USD
102666-498 102666-500 102666-502
4-Amino-5-[(4-fluorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(4-fluorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₉FN₄OS
MDL Number: MFCD03070533
CAS Number: 634157-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR