4-Amino-5-[4-(dimethylamino)phenyl]-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077482-500MG
077482-1G
077482-5G
102666-492EA
387.69
USD
102666-492
102666-494
102666-496
4-Amino-5-[4-(dimethylamino)phenyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[4-(dimethylamino)phenyl]-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₁₀H₁₃N₅S MW: 235.31 g/mol |
MDL Number:
MFCD07364330 CAS Number: 76457-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...