4-Amino-5-[(2,4-dichlorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077481-500MG
077481-1G
077481-5G
102666-486EA
258.46
USD
102666-486
102666-488
102666-490
4-Amino-5-[(2,4-dichlorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(2,4-dichlorophenoxy)methyl]-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₉H₈Cl₂N₄OS |
MDL Number:
MFCD03030345 CAS Number: 4413-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...