4-Amino-5-[(1-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077480-500MG 077480-1G 077480-5G
102666-480EA 258.46 USD
102666-480 102666-482 102666-484
4-Amino-5-[(1-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(1-naphthyloxy)methyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₁₂N₄OS
MDL Number: MFCD13370781
CAS Number: 137266-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR