4-Amino-5-[(phenylthio)methyl]-4H-1,2,4-triazole-3-thiol
Supplier: Matrix Scientific
077479-500MG
077479-1G
077479-5G
102666-474EA
387.69
USD
102666-474
102666-476
102666-478
4-Amino-5-[(phenylthio)methyl]-4H-1,2,4-triazole-3-thiol
4-Amino-5-[(phenylthio)methyl]-4H-1,2,4-triazole-3-thiol
Hazard: Irritant
Formula:
C₉H₁₀N₄S₂ |
MDL Number:
MFCD11868884 CAS Number: 178169-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...