4-Amino-5-(diphenylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077468-500MG 077468-1G
102666-418EA 258.46 USD
102666-418 102666-420
4-Amino-5-(diphenylmethyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(diphenylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₄N₄S
MDL Number: MFCD02672759
CAS Number: 93073-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR