4-Amino-3,4-dimethylpentanoic acid hydrochloride
Supplier: Matrix Scientific
077456-500MG
077456-1G
077456-5G
102666-346EA
646.15
USD
102666-346
102666-348
102666-350
4-Amino-3,4-dimethylpentanoic acid hydrochloride
4-Amino-3,4-dimethylpentanoic acid hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClNO₂ |
MDL Number:
MFCD20441442 CAS Number: 1401425-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...