4-(2-Ethoxyethyl)-1-phenylcyclohexanol

Supplier: Matrix Scientific

077439-500MG 077439-1G 077439-5G
102666-256EA 258.46 USD
102666-256 102666-258 102666-260
4-(2-Ethoxyethyl)-1-phenylcyclohexanol
4-(2-Ethoxyethyl)-1-phenylcyclohexanol

Hazard: Irritant

Formula: C₁₆H₂₄O₂
MDL Number: MFCD20441359
CAS Number: 1338494-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR