4-Methoxy-2,3,5-trimethylbenzaldehyde

Supplier: Matrix Scientific

077430-500MG 077430-1G
102666-204EA 646.15 USD
102666-204 102666-206
4-Methoxy-2,3,5-trimethylbenzaldehyde
4-Methoxy-2,3,5-trimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01652203
CAS Number: 59453-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR