2-(4-Nitro-1H-pyrazol-1-yl)acetamide

Supplier: Matrix Scientific

077425-500MG 077425-1G 077425-5G
102666-174EA 258.46 USD
102666-174 102666-176 102666-178
2-(4-Nitro-1H-pyrazol-1-yl)acetamide
2-(4-Nitro-1H-pyrazol-1-yl)acetamide

Hazard: Irritant

Formula: C₅H₆N₄O₃
MDL Number: MFCD00297185
CAS Number: 32407-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR