2-Chloro-3-(3,5-dimethyl-1H-pyrazol-1-yl)propanenitrile

Supplier: Matrix Scientific

077422-500MG 077422-1G 077422-5G
102666-156EA 387.69 USD
102666-156 102666-158 102666-160
2-Chloro-3-(3,5-dimethyl-1H-pyrazol-1-yl)propanenitrile
2-Chloro-3-(3,5-dimethyl-1H-pyrazol-1-yl)propanenitrile

Hazard: Irritant

Formula: C₈H₁₀ClN₃
MDL Number: MFCD20441351
CAS Number: 1338495-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR