Ethyl (3-nitro-1H-pyrazol-1-yl)acetate
Supplier: Matrix Scientific
077420-500MG
077420-1G
077420-5G
102666-144EA
387.69
USD
102666-144
102666-146
102666-148
Ethyl (3-nitro-1H-pyrazol-1-yl)acetate
Ethyl (3-nitro-1H-pyrazol-1-yl)acetate
Hazard: Irritant
Formula:
C₇H₉N₃O₄ |
MDL Number:
MFCD01622828 CAS Number: 102039-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...