3-[(2,5-Dimethylphenoxy)methyl]-4-methoxybenzoic acid
Supplier: Matrix Scientific
077409-500MG
077409-1G
077409-5G
102666-090EA
258.46
USD
102666-090
102666-092
102666-094
3-[(2,5-Dimethylphenoxy)methyl]-4-methoxybenzoic acid
3-[(2,5-Dimethylphenoxy)methyl]-4-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₇H₁₈O₄ |
MDL Number:
MFCD03421971 CAS Number: 438220-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...