5-[(2-Formylphenoxy)methyl]-2-furoic acid

Supplier: Matrix Scientific

077408-500MG 077408-1G 077408-5G
102666-084EA 387.69 USD
102666-084 102666-086 102666-088
5-[(2-Formylphenoxy)methyl]-2-furoic acid
5-[(2-Formylphenoxy)methyl]-2-furoic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₅
MDL Number: MFCD06653195
CAS Number: 881041-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR